missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Fluorescent Amidite Matrix Standards [6FAM™, TET, HEX, TAMRA™, and ROX™ dyes]

Product Code. 16365242 Shop All Applied Biosystems Products
Change view
Click to view available options
For Use With (Equipment):
310 Genetic Analyzer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
16365242 310 Genetic Analyzer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16365242 Supplier Applied Biosystems™ Supplier No. 401546

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

5 Tubes of dye labeled oligos, one tube for each dye

The ExoSAP-IT one-step solution provides a simple way to enzymatically purify a PCR reaction.

The Fluorescent Amidite Matrix Standards are used to generate the multicomponent matrix required when analyzing 6-FAM™, TET™, HEX™, TAMRA™ and ROX™-labeled DNA fragments.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 5 tubes of dye labeled oligos, one tube for each dye. Store the kit at 2-8°C. Do not freeze.
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Sequencing
Label or Dye FAM, HEX™, ROX, TAMRA™, TET™
Product Line FAM, HEX, ROX, TAMRA, TET
Product Type DNA Sequencing Reagents
Quantity 1 Ea.
Shipping Condition Wet Ice

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.