missing translation for 'onlineSavingsMsg'
Learn More

Fluoresceinamine, isomer I, 96%

Product Code. 15454599
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454599 5 g 5g
15444599 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454599 Supplier Thermo Scientific Alfa Aesar Supplier No. J62175.06
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A fluorescent labeling reagent | CAS: 3326-34-9 | C20H13NO5 | 347.33 g/mol

5-aminofluorescein is also used to prepare FITC Isomer I (CDX-F0011, CDX-F0020). It is a chemical used as molecular probe and important as fluorescent dye and for derivatives. Fluorescent labeling reagent for proteins. Used in the fluorescent antibody technique for rapid identification of pathogens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-aminofluorescein is also used to prepare FITC Isomer I (CDX-F0011, CDX-F0020). It is a chemical used as molecular probe and important as fluorescent dye and for derivatives. Fluorescent labeling reagent for proteins. Used in the fluorescent antibody technique for rapid identification of pathogens.

Solubility
Soluble in water (partly), methanol (10 mg/ml), and acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Light and Moisture Sensitive
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3326-34-9
Molecular Formula C20H13NO5
Molecular Weight (g/mol) 347.33
MDL Number MFCD00005052
InChI Key GZAJOEGTZDUSKS-UHFFFAOYSA-N
Synonym 5-Aminofluorescein
PubChem CID 76845
IUPAC Name 6-amino-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES NC1=CC=C2C(=C1)C(=O)OC21C2=CC=C(O)C=C2OC2=CC(O)=CC=C12

Specifications

Melting Point 223°C (decomposition)
Beilstein 48395
Color Brown to Red
Solubility Information Soluble in water (partly),methanol (10mg/ml),and acetone.
Quantity 5 g
Physical Form Powder
Formula Weight 347.32
Percent Purity 96%
Chemical Name or Material Fluoresceinamine, isomer I

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.