missing translation for 'onlineSavingsMsg'
Learn More

Fluoresceinamine, Isomer 1, TRC

Product Code. 30398989
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30398989 1 g
30400534 500 mg
30405868 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398989 Supplier Toronto Research Chemicals Supplier No. TRCF4853001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3326-34-9
Molecular Formula C20 H13 N O5
Molecular Weight (g/mol) 347.32
IUPAC Name 6-amino-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES Nc1ccc2c(c1)C(=O)OC23c4ccc(O)cc4Oc5cc(O)ccc35

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H13NO5/c21-10-1-4-14-13(7-10)19(24)26-20(14)15-5-2-11(22)8-17(15)25-18-9-12(23)3-6-16(18)20/h1-9,22-23H,21H2
Formula Weight 347.0794
Chemical Name or Material Fluoresceinamine, Isomer 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.