missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein Sodium Salt, indicator, 98.5 to 100.5%, Honeywell Fluka™
Indicator, 98.5-100.5%
£34.18 - £822.05
Chemical Identifiers
CAS | 518-47-8 |
---|---|
Molecular Formula | C20H10Na2O5 |
Molecular Weight (g/mol) | 376.28 |
MDL Number | MFCD00167039 |
InChI Key | NJDNXYGOVLYJHP-UHFFFAOYSA-L |
IUPAC Name | disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate |
SMILES | [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15682930
|
Honeywell-Fluka
28803-100G |
100 g | Glass Bottle |
£34.18
100g |
In Stock
Log in to see stock. |
||||
15692930
|
Honeywell-Fluka
28803-1KG |
1 kg | Glass Bottle |
£205.52
1kg |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||||
15602940
|
Honeywell-Fluka
28803-4X1KG |
4 x 1 kg | Glass Bottle |
£822.05
Pack of 4 |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
518-47-8 | |
376.28 | |
NJDNXYGOVLYJHP-UHFFFAOYSA-L | |
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12 |
C20H10Na2O5 | |
MFCD00167039 | |
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate |
Specifications
>300°C | |
C20H10Na2O5 | |
NONH for all modes of transport | |
NJDNXYGOVLYJHP-UHFFFAOYSA-L | |
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate | |
376.27g/mol | |
Indicator | |
Powder |
518-47-8 | |
MFCD00167039 | |
3833041 | |
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12 | |
376.28 | |
98.5 to 100.5% | |
Red | |
Fluorescein Sodium Salt |
Safety and Handling
missing translation for 'einecsNumber' : 208-253-0