missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein-Poly GAT

Product Code. 10368382
Click to view available options
Quantity:
1 mL
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 10368382

Brand: Invitrogen PV3611

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fluorescein-Poly GAT

A general fluorescein peptide substrate consisting of a polymer of glutamate, alanine, and tyrosine that is used to monitor tyrosine kinase activity.

For use in combination with the LanthaScreen™ Tb-pY20 Antibody (PV3552) to evaluate inhibitors in medium to high throughput screening applications.
TRUSTED_SUSTAINABILITY

Specifications

Amino Acid Sequence Fluorescein- Poly Glu:Ala:Tyr (6:3:1)
Content And Storage 30 μM Fluorescein Poly-GAT peptide substrate in 10 mM HEPES (pH 7.5), 137 mM NaCl, and 2.7 mM KCl (HEPES buffered saline). Store at -20°C and protect from light.
Detection Method Fluorescence
Quantity 1 mL
Substrate Fluorescein-Poly GAT
Substrate Properties Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Tyrosine Kinases
Label or Dye FITC (Fluorescein)
Research Category Signal Transduction
Shipping Condition Dry Ice
Validated Kinases ABL1, ABL2, AXL, BLK, BMX, BTK, CSF1R, CSK, DDR2, EGFR, EPHA1, EPHA2, EPHA3, EPHA4, EPHA5, EPHA7, EPHA8, EPHB1, EPHB2, EPHB3, EPHB4, ERBB2, ERBB4, FER, FGFR1, FGFR2, FGFR3, FGFR4, FGR, FLT1, FLT3, FLT4, FRK, FYN, HCK, IGF1R, INSR, KDR, KIT, LCK, LTK, LYNA, LYNB, MATK, MERTK, MET, MST1R, MUSK, NTRK1, NTRK2, NTRK3, PDGFRA, PDGFRB, PTK2, PTK2B, PTK6, RET, ROS1, SRC, SRCN1, SRMS, SYK, TEC, TEK, TNK2, TTK, TYRO3, YES1, ZAP70
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.