missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein Isothiocyanate (>85%), TRC

Product Code. 30404467
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30404467 100 mg
30389024 250 mg
30402367 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30404467 Supplier Toronto Research Chemicals Supplier No. TRCF485715100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27072-45-3
Molecular Formula C21H11NO5S
Molecular Weight (g/mol) 389.38
Synonym FITC, Isothiocyanato Fluorescein, 3',6'-Dihydroxy-5(or 6)-isothiocyanatospiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one Incompletely Defined Substance, 5(6)-FITC
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O.CN=C=S

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H11NO5S/c23-12-2-5-16-18(8-12)26-19-9-13(24)3-6-17(19)21(16)15-4-1-11(22-10-28)7-14(15)20(25)27-21/h1-9,23-24H
Chemical Name or Material Fluorescein Isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.