missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluorescein, disodium salt, pure

Product Code. 10206090 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic drum
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10206090

Brand: Thermo Scientific Chemicals 173245000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 518-47-8 | C20H10Na2O5 | 376.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 518-47-8
Molecular Formula C20H10Na2O5
Molecular Weight (g/mol) 376.28
MDL Number MFCD00167039
InChI Key NJDNXYGOVLYJHP-UHFFFAOYSA-L
IUPAC Name disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12

Specifications

Absorbance 1700 (at lambda max.) min.
Melting Point 320.0°C
Absorption Ratio 1.2 to 1.7
Beilstein 19, V,6, 456
Merck Index 15, 4192
Synonym Acid Yellow 73, Fluorescein, water soluble, Uranine, C.I. 45350
Solubility Information Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
Infrared Spectrum Authentic
Lambda Max 488 to 491nm (in buffer pH 8)
Formula Weight 376.27
Loss on Drying 10% (105°C) max.
Grade Pure
Packaging Glass Bottle
Color Brown to Orange/Red or Brown/Green
Quantity 500 g
Physical Form Powder
Chemical Name or Material Fluorescein, disodium salt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.