missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein diacetate, 97%, Alfa Aesar™

Product Code. 11485927
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485927 25 g 25g
11475927 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485927 Supplier Thermo Scientific Alfa Aesar Supplier No. B24466.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Applications
Fluorescein diacetate stains living cells green, and is a substrate for esterases. It is suitable for use in cell viability assays. Removal of the acetate groups by cellular esterases results in fluorescence in cells with intact membranes. It is used in studies on intracellular interactions and membrane permeability.

Solubility
Soluble in chloroform, DMSO, ethanol, and water.

Notes
Store at -20°C. Store in a cool, dry place in tightly closed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-09-8
Molecular Formula C24H16O7
Molecular Weight (g/mol) 416.385
MDL Number MFCD00005062
InChI Key CHADEQDQBURGHL-UHFFFAOYSA-N
Synonym fluorescein diacetate, 3,6-diacetoxyfluoran, diacetylfluorescein, di-o-acetylfluorescein, fluorescein, diacetate, 3',6'-diacetylfluorescein, unii-yl39r93pre, yl39r93pre, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 3',6'-bis acetyloxy, 3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-3',6'-diyl diacetate
PubChem CID 65047
IUPAC Name (6'-acetyloxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-3'-yl) acetate
SMILES CC(=O)OC1=CC2=C(C=C1)C3(C4=C(O2)C=C(C=C4)OC(=O)C)C5=CC=CC=C5C(=O)O3

Specifications

Melting Point 200°C to 203°C
Assay Percent Range 0.97
Quantity 25 g
Beilstein 62575
Formula Weight 416.39
Percent Purity 97%
Chemical Name or Material Fluorescein diacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.