missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB600-493
This item is not returnable.
View return policy
Description
Fluorescein Polyclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount.
Specifications
| Fluorescein | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Fluorescein | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry | |
| 1 mg/ml | |
| Western Blot 1:500-1:3000, Flow Cytometry, ELISA 1:5000-1:25000, Immunohistochemistry 1:250-1:1500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount | |
| FITC, fluorescein isothyocianate | |
| Fluorescein conjugated to Goat IgG | |
| 0.2 mg | |
| Primary | |
| Non-species specific | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction