missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein Antibody (4-4-20 (enhanced)), DyLight 405, Novus Biologicals™

Product Code. 18392131 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392131 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392131 Supplier Novus Biologicals Supplier No. NBP281026V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Fluorescein Monoclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Western Blot, Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fluorescein
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 4-4-20 (enhanced)
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias FITC, fluorescein isothyocianate
Host Species Rat
Immunogen Fluorescein I-bovine serum albumin (FlI-BSA).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Fluorescein labelled proteins.
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.