missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein 6-Isothiocyanate, Isomer 2, 95%, TRC

Product Code. 30396634
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396634 1 g
30408656 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396634 Supplier Toronto Research Chemicals Supplier No. TRCF4857051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18861-78-4
Molecular Formula C21 H11 N O5 S
Molecular Weight (g/mol) 389.38
Synonym Fluorescein 6-Isothiocyanate, Isomer 2
IUPAC Name 3',6'-dihydroxy-5-isothiocyanatospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES Oc1ccc2c(Oc3cc(O)ccc3C24OC(=O)c5ccc(cc45)N=C=S)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H11NO5S/c23-12-2-5-15-18(8-12)26-19-9-13(24)3-6-16(19)21(15)17-7-11(22-10-28)1-4-14(17)20(25)27-21/h1-9,23-24H
Formula Weight 389.0358
Chemical Name or Material Fluorescein 6-Isothiocyanate, Isomer 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.