Learn More
Fluorescamine, Thermo Scientific Chemicals
For fluorescent detection of amino acids, peptides and proteins
£25.20 - £275.00
Chemical Identifiers
CAS | 38183-12-9 |
---|---|
Molecular Formula | C17H10O4 |
Molecular Weight (g/mol) | 278.26 |
MDL Number | MFCD00005928 |
InChI Key | ZFKJVJIDPQDDFY-UHFFFAOYNA-N |
Synonym | fluorescamine, fluram, spiro furan-2 3h ,1' 3'h-isobenzofuran-3,3'-dione, 4-phenyl, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dione, 4-phenylspiro furan-2 3h ,1'-phthalan-3,3'-dione, 4-phenylspiro furan-2 3h-1'-3'-h-isobenzofuran-3,3'-dione, zfkjvjidpqddfy-uhfffaoysa, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dion |
PubChem CID | 37927 |
SMILES | O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11370368
|
Thermo Scientific Alfa Aesar
043749.MB |
25 mg |
£25.20
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11380368
|
Thermo Scientific Alfa Aesar
043749.MC |
100 mg |
£81.70
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11360368
|
Thermo Scientific Alfa Aesar
043749.ME |
500 mg |
£275.00
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For fluorescent detection of amino acids, peptides and proteins.Fluorescamine plays an important role as a reagent for the detection of amines, peptides and proteins. It is also used as a substrate for measuring protease activity. It is utilized in the detection and quantitation of residual aminopenicillins and sulfonamides in honey by HPLC. It finds application in the determination of lisinopril in human plasma and urine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
38183-12-9 | |
278.26 | |
ZFKJVJIDPQDDFY-UHFFFAOYNA-N | |
37927 |
C17H10O4 | |
MFCD00005928 | |
fluorescamine, fluram, spiro furan-2 3h ,1' 3'h-isobenzofuran-3,3'-dione, 4-phenyl, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dione, 4-phenylspiro furan-2 3h ,1'-phthalan-3,3'-dione, 4-phenylspiro furan-2 3h-1'-3'-h-isobenzofuran-3,3'-dione, zfkjvjidpqddfy-uhfffaoysa, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dion | |
O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12 |
Specifications
38183-12-9 | |
White to Cream | |
MFCD00005928 | |
fluorescamine, fluram, spiro furan-2 3h ,1' 3'h-isobenzofuran-3,3'-dione, 4-phenyl, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dione, 4-phenylspiro furan-2 3h ,1'-phthalan-3,3'-dione, 4-phenylspiro furan-2 3h-1'-3'-h-isobenzofuran-3,3'-dione, zfkjvjidpqddfy-uhfffaoysa, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dion | |
ZFKJVJIDPQDDFY-UHFFFAOYNA-N | |
4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione | |
37927 | |
Powder |
155°C to 158°C | |
C17H10O4 | |
14,4158 | |
Soluble in acetone,ethanol,chloroform,dimethylsulfoxide and acetonitrile. Slightly soluble in water. | |
O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12 | |
278.26 | |
278.27 | |
Fluorescamine |
Safety and Handling
missing translation for 'einecsNumber' : 253-814-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only