missing translation for 'onlineSavingsMsg'
Learn More

Fluorene, TRC

Product Code. 30405067
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30405067 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405067 Supplier Toronto Research Chemicals Supplier No. TRCF4620001G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 86-73-7
Molecular Formula C13 H10
Quantity 1 g
Synonym Fluorene, 9H-Fluorene, Fluorene (8CI), 2,2'-Methylenebiphenyl, Diphenylenemethane, Methane, diphenylene-, NSC 6787, o-Biphenylenemethane
InChI Formula InChI=1S/C13H10/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)12/h1-8H,9H2
SMILES C1c2ccccc2c3ccccc13
IUPAC Name 9H-fluorene
Molecular Weight (g/mol) 166.22
Formula Weight 166.0783
Chemical Name or Material Fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.