missing translation for 'onlineSavingsMsg'
Learn More

Fluorene-D10, TRC

Product Code. 30389633
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389633 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30389633 Supplier Toronto Research Chemicals Supplier No. TRCF462003100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluorene D10
CAS 81103-79-9
Molecular Formula C13 D10
Quantity 100 mg
Synonym 9H-Fluorene-1,2,3,4,5,6,7,8,9,9-d10, Fluorene D10, Perdeuterofluorene
InChI Formula InChI=1S/C13H10/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)12/h1-8H,9H2/i1D,2D,3D,4D,5D,6D,7D,8D,9D2
SMILES [2H]c1c([2H])c([2H])c2c(c1[2H])c3c([2H])c([2H])c([2H])c([2H])c3C2([2H])[2H]
IUPAC Name 1,2,3,4,5,6,7,8,9,9-decadeuteriofluorene
Molecular Weight (g/mol) 176.2801
Formula Weight 176.141
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.