missing translation for 'onlineSavingsMsg'
Learn More

Fluorene, 98+%, Thermo Scientific Chemicals

Catalog Number 11391517
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
86-73-7
C13H10
166.22
MFCD00001111
NIHNNTQXNPWCJQ-UHFFFAOYSA-N
fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene
6853
CHEBI:28266
9H-fluorene
C1C2=CC=CC=C2C2=CC=CC=C12
This item is not returnable. View return policy
86-73-7
C13H10
166.22
MFCD00001111
NIHNNTQXNPWCJQ-UHFFFAOYSA-N
fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene
6853
CHEBI:28266
9H-fluorene
C1C2=CC=CC=C2C2=CC=CC=C12

Fluorene is used in the preparation of dyes such as 2,7-diiodofluorene, 2-aminofluorene and 3,6-bis-(dimethylamino)fluorine as well as resins. It acts as a precursor to other fluorene compounds and to prepare fluorenone. It is utilized in the study of the extraction of specific semiconducting single-wall carbon nanotubes. Its polymer, polyfluorene finds application in electrically conductive and electroluminescent as well as in solar cells. In addition to this, it is used as a luminophore inorganic light-emitting diodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorene is used in the preparation of dyes such as 2,7-diiodofluorene, 2-aminofluorene and 3,6-bis-(dimethylamino)fluorine as well as resins. It acts as a precursor to other fluorene compounds and to prepare fluorenone. It is utilized in the study of the extraction of specific semiconducting single-wall carbon nanotubes. Its polymer, polyfluorene finds application in electrically conductive and electroluminescent as well as in solar cells. In addition to this, it is used as a luminophore inorganic light-emitting diodes

Solubility
Soluble in hot alcohol, ether, benzene and glacial acetic acid. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 113°C to 117°C
Density 1.202
Boiling Point 295°C
Flash Point 151°C (303°F)
Quantity 25 g
UN Number UN3077
Beilstein 1363491
Merck Index 14,4155
Solubility Information Soluble in hot alcohol,ether,benzene and glacial acetic acid. Insoluble in water.
Formula Weight 166.22
Percent Purity ≥98%
Chemical Name or Material Fluorene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'