missing translation for 'onlineSavingsMsg'
Learn More

Fluorene-9-acetic acid, 99+%

Product Code. 11473048
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473048 1 g 1g
11483048 5 g 5g
11493048 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473048 Supplier Thermo Scientific Alfa Aesar Supplier No. A18772.03
  • ONLINE EXCLUSIVE PRICE
    £45.80
    £41.65 / 1g
    Save £4.15
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6284-80-6 | C15H12O2 | 224.259 g/mol

It was used in preparation of pyrokinin class of insect neuropeptides. It was also used in preparation of N-(9-fluoreneacetyl)-L-leucine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It was used in preparation of pyrokinin class of insect neuropeptides. It was also used in preparation of N-(9-fluoreneacetyl)-L-leucine.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6284-80-6
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00013262
InChI Key WFSMJMTYIMFHPV-UHFFFAOYSA-N
Synonym 9-fluoreneacetic acid, fluorene-9-acetic acid, 9h-fluorene-9-acetic acid, 2-9h-fluoren-9-yl acetic acid, 9h-fluorene-9-aceticacid, 9h-fluoren-9-ylacetic acid, chembl44381, 2-fluoren-9-ylacetic acid, 9-fluoreneacetate, 9-fluorene acetic acid
PubChem CID 94852
IUPAC Name 2-(9H-fluoren-9-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)CC(=O)O

Specifications

Melting Point 133°C to 135°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 224.26
Percent Purity ≥99.5%
Chemical Name or Material Fluorene-9-acetic acid
compliance-icons
Product Identifier
  • Fluorene-9-acetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 9H-Fluorene-9-aceticacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.