missing translation for 'onlineSavingsMsg'
Learn More

Fluorene-4-carboxylic acid, 96%

Product Code. 11334494
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334494 250 mg 250mg
11314494 1 g 1g
11324494 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334494 Supplier Thermo Scientific Alfa Aesar Supplier No. L01164.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6954-55-8 | C14H10O2 | 210.232 g/mol

Fluorene-4-carboxylic acid is used in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorene-4-carboxylic acid is used in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6954-55-8
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.232
MDL Number MFCD00001112
InChI Key WJNBLUOXTBTGMB-UHFFFAOYSA-N
Synonym fluorene-4-carboxylic acid, 4-fluorenecarboxylic acid, maybridge1_002107, 9h-fluorene-4-carboxylicacid, fluorene-4-carboxylic acid, technical grade
PubChem CID 81402
IUPAC Name 9H-fluorene-4-carboxylic acid
SMILES C1C2=CC=CC=C2C3=C(C=CC=C31)C(=O)O

Specifications

Melting Point 191°C to 192°C
Quantity 250 mg
Beilstein 2050637
Solubility Information Insoluble in water.
Formula Weight 210.23
Percent Purity 96%
Chemical Name or Material Fluorene-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.