missing translation for 'onlineSavingsMsg'
Learn More

Fluorene-13C6, TRC

Product Code. 30394888
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394888 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394888 Supplier Toronto Research Chemicals Supplier No. TRCF4620021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluorene-13C6
CAS 1189497-69-5
Molecular Formula 13C6 C7 H10
Quantity 1 mg
Synonym Fluorene 13C6
InChI Formula InChI=1S/C13H10/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)12/h1-8H,9H2/i1+1,3+1,5+1,7+1,10+1,12+1
SMILES C1[13c]2[13cH][13cH][13cH][13cH][13c]2c3ccccc13
IUPAC Name 9H-fluorene
Molecular Weight (g/mol) 172.17
Formula Weight 172.0984
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.