missing translation for 'onlineSavingsMsg'
Få mere at vide

Fluoren-3-ol-d9 (Major), TRC

Produktkode 30392722
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
10 mg
Förpackningsstorlek
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Quantity unitSize
30392722 10 mg 10mg
30404564 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Produktkode 30392722 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCF46246210MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Fluoren-3-ol-d9
Molecular Formula C13HD9O
Quantity 10 mg
Synonym 3-Hydroxyfluorene-d9, NSC 51318-d9, 9H-Fluoren-3-ol-d9, Fluoren-1,2,4,5,6,7,8,9,9-d9-3-ol
InChI Formula InChI=1S/C13H10O/c14-11-6-5-10-7-9-3-1-2-4-12(9)13(10)8-11/h1-6,8,14H,7H2/i1D,2D,3D,4D,5D,6D,7D2,8D
SMILES OC1=C([2H])C2=C(C([2H])=C1[2H])C([2H])([2H])C3=C([2H])C([2H])=C([2H])C([2H])=C32
Molecular Weight (g/mol) 191.27
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.