missing translation for 'onlineSavingsMsg'
Learn More

Fluoren-1-ol, TRC

Product Code. 30402739
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402739 5 mg
30405374 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402739 Supplier Toronto Research Chemicals Supplier No. TRCF4624505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6344-61-2
Molecular Formula C13 H10 O
Molecular Weight (g/mol) 182.22
Synonym 9H-Fluoren-1-ol (9CI, ACI), Fluoren-1-ol (6CI, 7CI, 8CI), 1-Hydroxyfluorene, NSC 51310
IUPAC Name 9H-fluoren-1-ol
SMILES Oc1cccc2c1Cc3ccccc23

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H10O/c14-13-7-3-6-11-10-5-2-1-4-9(10)8-12(11)13/h1-7,14H,8H2
Formula Weight 182.0732
Chemical Name or Material Fluoren-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.