Learn More
Fluoranthene, 98%, Thermo Scientific Chemicals
£36.00 - £301.00
Chemical Identifiers
CAS | 206-44-0 |
---|---|
Molecular Formula | C16H10 |
Molecular Weight (g/mol) | 202.26 |
MDL Number | MFCD00001184 |
InChI Key | GVEPBJHOBDJJJI-UHFFFAOYSA-N |
Synonym | idryl, 1,2-benzacenaphthene, benzo jk fluorene, rcra waste number u120, 1,2-1,8-naphthylene benzene, benzene, 1,2-1,8-naphthalenediyl, 1,2-1,8-naphthalenediyl benzene, benzene, 1,2-1,8-naphthylene, benzo j,k fluorene |
PubChem CID | 9154 |
ChEBI | CHEBI:33083 |
IUPAC Name | fluoranthene |
SMILES | C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10182380
|
Thermo Scientific Acros
119170250 |
25 g | Glass bottle |
£36.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10102390
|
Thermo Scientific Acros
119171000 |
100 g | Glass bottle |
£95.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10646112
|
Thermo Scientific Acros
119175000 |
500 g | Glass bottle |
£301.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
206-44-0 | |
202.26 | |
GVEPBJHOBDJJJI-UHFFFAOYSA-N | |
9154 | |
fluoranthene |
C16H10 | |
MFCD00001184 | |
idryl, 1,2-benzacenaphthene, benzo jk fluorene, rcra waste number u120, 1,2-1,8-naphthylene benzene, benzene, 1,2-1,8-naphthalenediyl, 1,2-1,8-naphthalenediyl benzene, benzene, 1,2-1,8-naphthylene, benzo j,k fluorene | |
CHEBI:33083 | |
C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13 |
Specifications
206-44-0 | |
Beige-Gray to Green-Yellow or Yellow | |
>100°C | |
97.5% min. (GC) | |
MFCD00001184 | |
idryl, 1,2-benzacenaphthene, benzo jk fluorene, rcra waste number u120, 1,2-1,8-naphthylene benzene, benzene, 1,2-1,8-naphthalenediyl, 1,2-1,8-naphthalenediyl benzene, benzene, 1,2-1,8-naphthylene, benzo j,k fluorene | |
GVEPBJHOBDJJJI-UHFFFAOYSA-N | |
fluoranthene | |
9154 | |
202.26 | |
Crystalline Powder, Crystals or Chunks |
109.0°C to 111.0°C | |
384.0°C | |
Authentic | |
C16H10 | |
05, I, 344 | |
Solubility in water: insoluble. Other solubilities: soluble in ether, acetone and benzene | |
C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13 | |
202.26 | |
CHEBI:33083 | |
98% | |
Fluoranthene, 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not eat,drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release t
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 205-912-4
missing translation for 'rtecsNumber' : LL4025000
missing translation for 'tsca' : TSCA
- Fluoranthene
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only