missing translation for 'onlineSavingsMsg'
Learn More

Fluopyram-benzamide, Dr. Ehrenstorfer

Codice prodotto. 30323621
Cambia vista
Click to view available options
Quantity:
100 mg
Dimensione della confezione:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
30323621 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30323621 Fornitore Dr. Ehrenstorfer N. del fornitore DREC13743100
 Restricted Item
Estimated Shipment: 30-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluopyram-benzamide
CAS 360-64-5
Analyte or Component Names Fluopyram-benzamide
Shipping Condition Room Temperature
Molecular Formula C8 H6 F3 N O
Certifications/Compliance ISO 17034
Synonym o-Toluamide, α,α,α-trifluoro- (6CI,8CI), 2-(Trifluoromethyl)benzamide
InChI Formula InChI=1S/C8H6F3NO/c9-8(10,11)6-4-2-1-3-5(6)7(12)13/h1-4H,(H2,12,13)
Solution Type Neat
SMILES NC(=O)c1ccccc1C(F)(F)F
IUPAC Name 2-(trifluoromethyl)benzamide
Industry Type Environmental Testing
Molecular Weight (g/mol) 189.13
Quantity 100 mg
Formula Weight 189.0401 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.