missing translation for 'onlineSavingsMsg'
Learn More

Fluocinonide, Mikromol™

Product Code. 30052398
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30052398

Brand: Mikromol MM3001.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Fluocinonide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluocinolone acetonide acetate
CAS 356-12-7
Analyte or Component Names Fluocinolone acetonide acetate
Shipping Condition Room Temperature
Molecular Formula C26 H32 F2 O7
Certifications/Compliance ISO 17034
Synonym (6alpha,11beta,16alpha)-21-(Acetyloxy)-6,9-difluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, 6alpha,9-Difluoro-11beta,16alpha,17,21-tetrahydroxypregna-1,4-diene-3,20-dione 21-acetate cyclic 16,17-acetal with acetone, 2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole pregna-1,4-diene-3,20-dione deriv., Biscosal, Dermaplus, Flucetonide, Flucinar, Fluocinolide, Fluocinolide acetate, Fluocinolone acetonide 21-acetate, Fluocinolone acetonide acetate, Fluocinonide, Lidex, Lidex E, Metosyn, NSC 101791, Straderm, Topsym, Topsymin, Topsyn, Topsyne, Vanos, 6α,9-Difluoro-11β-hydroxy-16α,17-(1-methylethylidenedioxy)-3,20-dioxopregna-1,4-dien-21-yl Acetate
InChI Formula InChI=1S/C26H32F2O7/c1-13(29)33-12-20(32)26-21(34-22(2,3)35-26)10-15-16-9-18(27)17-8-14(30)6-7-23(17,4)25(16,28)19(31)11-24(15,26)5/h6-8,15-16,18-19,21,31H,9-12H2,1-5H3/t15-,16-,18-,19-,21+,23-,24-,25-,26+/m0/s1
SMILES CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]23C
Molecular Weight (g/mol) 494.52
Quantity 250 mg
Formula Weight 494.2116 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.