missing translation for 'onlineSavingsMsg'
Learn More

Fluocinolone Acetonide, TRC

Product Code. 30398707
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398707 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398707 Supplier Toronto Research Chemicals Supplier No. TRCF4558002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluocinolone Acetonide
CAS 67-73-2
Purity Grade Notes HPLC
Molecular Formula C24 H30 F2 O6
Quantity 2.5 g
Synonym Fluocinolone acetonide, 6α,9-Difluoro-11β,21-dihydroxy-16α,17-(1-methylethylidenedioxy)pregna-1,4-diene-3,20-dione, Pregna-1,4-diene-3,20-dione, 6,9-difluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (6α,11β,16α)-, Pregna-1,4-diene-3,20-dione, 6α,9-difluoro-11β,16α,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone (6CI,8CI), (6α,11β,16α)-6,9-Difluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, 2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregna-1,4-diene-3,20-dione deriv., 16α,17α-Isopropylidene-6α,9α-difluoro-1,4-pregnadiene-11β,16α,17α,21-tetrol-3,20-dione, 6α,9-Difluoro-11β,16α,17,21-tetrahydroxypregna-1,4-diene-3,20-dione cyclic 16,17 acetal with acetone, 6α,9-Difluoro-11β,21-dihydroxy-16α,17α-(isopropylidenedioxy)pregna-1,4-diene-3,20-dione, 6α,9α-Difluoro-11β,16α,17α,21-tetrahydroxypregna-1,4-diene-3,20-dione 16,17-acetonide, 6α,9α-Difluoro-16α,17α-isopropylidenedioxy-Δ1-hydrocortisone, 6α,9α-Difluoro-16α-hydroxyprednisolone 16,17-acetonide, 6α-Fluorotriamcinolone acetonide, Coriphate, Cortiplastol, Dermalar, Dermatin, Dermatin (steroid), Fluocinolone 16,17-acetonide, Fluocinolone 16α,17α-acetonide, Fluonid, Fluovitef, Fluovitif, Fluvean, Fluzon, Iluvien, Jellin, Localyn, Localyn Syntex, NSC 92339, Percutina, Prodermin, RS 1401AT, Radiocin, Retisert, Sinaflan, Sinalar, Synaflan, Synalar, Synamol, Synandone, Synandrone, Synemol, Synotic, Synsac
InChI Formula InChI=1S/C24H30F2O6/c1-20(2)31-19-9-13-14-8-16(25)15-7-12(28)5-6-21(15,3)23(14,26)17(29)10-22(13,4)24(19,32-20)18(30)11-27/h5-7,13-14,16-17,19,27,29H,8-11H2,1-4H3/t13-,14-,16-,17-,19+,21-,22-,23-,24+/m0/s1
SMILES CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
IUPAC Name (1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-16-one
Molecular Weight (g/mol) 452.49
Formula Weight 452.201
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.