missing translation for 'onlineSavingsMsg'
Learn More

Fluocinolone Acetonide-13C3, TRC

Product Code. 30389632
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389632 10 mg -
30393169 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389632 Supplier Toronto Research Chemicals Supplier No. TRCF45580210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1262192-25-5
Molecular Formula C2113C3H30F2O6
Molecular Weight (g/mol) 455.47
Synonym (6α,11β,16α)-6,9-Difluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-pregna-1,4-diene-3,20-dione-13C3, 6α-Fluorotriamcinolone Acetonide-13C3, Fluonid-13C3, Fluzon-13C3, Radiocin-13C3, Synandone-13C3, NSC 92339-13C3
SMILES [H][C@]12[C@]3([H])C[C@@H](C4=CC(C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]1(C)[C@@]5(C(CO)=O)C(O[13C]([13CH3])([13CH3])O5)C2)=O)F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H30F2O6/c1-20(2)31-19-9-13-14-8-16(25)15-7-12(28)5-6-21(15,3)23(14,26)17(29)10-22(13,4)24(19,32-20)18(30)11-27/h5-7,13-14,16-17,19,27,29H,8-11H2,1-4H3
Chemical Name or Material Fluocinolone Acetonide-13C3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.