missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Fluo-4 AM

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Fluo-4 AM is a cell-permeable, fluorescent Ca2+ indicator (Kd Ca2+ = 345 nM). Displays no resting signal and 100-fold increase in emission intensity upon Ca2+ binding. Excitation/emission λ 494/506 nm.
Specifications
Specifications
| Chemical Name or Material | N-[4-[6-[(Acetyloxy)methoxy]-2,7-difluoro-3-oxo-3H-xanthen-9-yl]-2-[2-[2-[bis[2-[(acetyloxy)methoxy]-2-oxoethyl]amino]-5-methylphenoxy]ethoxy]phenyl]-N-[2-[(acetyloxy)methoxy]-2-oxoethyl]glycine (acetyloxy)methyl ester |
| CAS | 273221-67-3 |
| Quantity | 1 mg |
| Target | Calcium Signaling Probes and Ca2+ Indicator Dyes |
| Molecular Formula | C51H50F2N2O23 |
| Formula Weight | 1096.95 |
| Purity | 95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction