missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flunixin Meglumine, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Flunixin Meglumine
Specifications
Specifications
| Chemical Name or Material | Flunixin meglumine |
| CAS | 42461-84-7 |
| Analyte or Component Names | Flunixin meglumine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H11 F3 N2 O2 . C7 H17 N O5 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Flunixin Meglumine |
| InChI Formula | InChI=1S/C14H11F3N2O2.C7H17NO5/c1-8-10(14(15,16)17)5-2-6-11(8)19-12-9(13(20)21)4-3-7-18-12;1-8-2-4(10)6(12)7(13)5(11)3-9/h2-7H,1H3,(H,18,19)(H,20,21);4-13H,2-3H2,1H3/t;4-,5+,6+,7+/m.0/s1 |
| SMILES | CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.Cc1c(Nc2ncccc2C(=O)O)cccc1C(F)(F)F |
| IUPAC Name | (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol;2-[2-methyl-3-(trifluoromethyl)anilino]pyridine-3-carboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction