missing translation for 'onlineSavingsMsg'
Learn More

Flunixin Meglumine EP Impurity A (Niflumic Acid EP Impurity A), Mikromol™

Product Code. 30461225
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461225 4 x 25 mg -
30050157 25 mg 25mg
17933146 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30461225 Supplier Mikromol Supplier No. MM0915.0100254AMP
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Flunixin Meglumine EP Impurity A (Niflumic Acid EP Impurity A), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloronicotinic acid
CAS 2942-59-8
Analyte or Component Names 2-Chloronicotinic acid
Shipping Condition Room Temperature
Molecular Formula C6 H4 Cl N O2
Certifications/Compliance ISO 17025
Synonym 2-Chloropyridine-3-carboxylic Acid, Flunixin Meglumine Imp. A (EP), Niflumic Acid Imp. A (EP), Flunixin USP Related Compound A, Flunixin USP RC A, 2-Chloropyridine-3-carboxylic Acid, 2-Chloronicotinic Acid, Flunixin Related Compound A, Flunixin Meglumine Impurity A, Niflumic Acid Impurity A, Flunixin Impurity A
InChI Formula InChI=1S/C6H4ClNO2/c7-5-4(6(9)10)2-1-3-8-5/h1-3H,(H,9,10)
Solution Type Neat
SMILES OC(=O)c1cccnc1Cl
IUPAC Name 2-chloropyridine-3-carboxylic acid
Molecular Weight (g/mol) 157.55
Quantity 4 x 25 mg
Formula Weight 156.9931 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.