missing translation for 'onlineSavingsMsg'
Learn More

Flunisolide, TRC

Product Code. 30402545
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402545 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402545 Supplier Toronto Research Chemicals Supplier No. TRCF50048510MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flunisolide Hemihydrate
CAS 77326-96-6
Molecular Formula 2 C24 H31 F O6 . H2 O
Quantity 10 mg
Synonym (6α,11β,16α)-6-Fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione Hydrate (2:1), Flunisolide Hemihydrate
InChI Formula InChI=1S/2C24H31FO6.H2O/c2*1-21(2)30-19-9-14-13-8-16(25)15-7-12(27)5-6-22(15,3)20(13)17(28)10-23(14,4)24(19,31-21)18(29)11-26;/h2*5-7,13-14,16-17,19-20,26,28H,8-11H2,1-4H3;1H2/t2*13-,14-,16-,17-,19+,20+,22-,23-,24+;/m00./s1
SMILES O.CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO.CC6(C)O[C@@H]7C[C@H]8[C@@H]9C[C@H](F)C%10=CC(=O)C=C[C@]%10(C)[C@H]9[C@@H](O)C[C@]8(C)[C@@]7(O6)C(=O)CO
IUPAC Name (6α,11β,16α)-6-Fluoro-11,21-dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione hemihydrate
Molecular Weight (g/mol) 887.01
Formula Weight 886.4315
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.