missing translation for 'onlineSavingsMsg'
Learn More

Flunarizine Dihydrochloride, Mikromol™

Product Code. 17901136
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17901136 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17901136 Supplier Mikromol Supplier No. MM0175.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Flunarizine Dihydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flunarizine dihydrochloride
CAS 30484-77-6
Analyte or Component Names Flunarizine dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C26 H26 F2 N2 . 2 Cl H
Certifications/Compliance ISO 17034
Synonym Piperazine, 1-[bis(4-fluorophenyl)methyl]-4-[(2E)-3-phenyl-2-propen-1-yl]-, hydrochloride (1:2), Piperazine, 1-[bis(4-fluorophenyl)methyl]-4-(3-phenyl-2-propenyl)-, dihydrochloride, (E)-, Piperazine, 1-[bis(4-fluorophenyl)methyl]-4-[(2E)-3-phenyl-2-propenyl]-, dihydrochloride (9CI), Piperazine, 1-[bis(p-fluorophenyl)methyl]-4-cinnamyl-, dihydrochloride, (E)- (8CI), (E)-1-[Bis(4-fluorophenyl)methyl]-4-cinnamylpiperazine dihydrochloride, Dinaplex, Flugeral, Flunagen, Flunarizine dihydrochloride, Flunarizine hydrochloride, Flunarl, Fluxarten, Gradient, Issium, Mondus, R 14950
InChI Formula InChI=1S/C26H26F2N2.2ClH/c27-24-12-8-22(9-13-24)26(23-10-14-25(28)15-11-23)30-19-17-29(18-20-30)16-4-7-21-5-2-1-3-6-21;;/h1-15,26H,16-20H2;2*1H/b7-4+;;
SMILES Cl.Cl.Fc1ccc(cc1)C(N2CCN(C\C=C\c3ccccc3)CC2)c4ccc(F)cc4
IUPAC Name 1-[bis(4-fluorophenyl)methyl]-4-[(E)-3-phenylprop-2-enyl]piperazine;dihydrochloride
Molecular Weight (g/mol) 477.42
Quantity 500 mg
Formula Weight 476.1598 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.