missing translation for 'onlineSavingsMsg'
Learn More

Flunarizine dihydrochloride, MedChemExpress

Product Code. 16422116
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422116 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422116 Supplier Medchem Express Supplier No. HYB0358A500mg
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Flunarizine dihydrochloride is a potent dual Na+/Ca2+ channel (T-type) blocker. Flunarizine dihydrochloride is a D2 dopamine receptor antagonist. Flunarizine dihydrochloride, a diphenylpiperazine derivative, has the potential for peripheral vessels dilator and migraine prophylaxis.

Flunarizine dihydrochloride is a potent dual Na+/Ca2+ channel (T-type) blocker. Flunarizine dihydrochloride is a D2 dopamine receptor antagonist. Flunarizine dihydrochloride, a diphenylpiperazine derivative, has the potential for peripheral vessels dilator and migraine prophylaxis.

Specifications

Chemical Name or Material Flunarizine dihydrochloride
CAS 30484-77-6
Color White
Purity Grade Notes Research
Molecular Formula C26H28Cl2F2N2
Quantity 500 mg
Solubility Information DMSO : 50 mg/mL (104.73 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.09 mM; Need ultrasonic)
SMILES FC1=CC=C(C(N2CCN(C/C=C/C3=CC=CC=C3)CC2)C4=CC=C(F)C=C4)C=C1.Cl.Cl
Molecular Weight (g/mol) 477.42
Formula Weight 477.42
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.