missing translation for 'onlineSavingsMsg'
Learn More

Flunarizine dihydrochloride, 99%

Product Code. 15465259
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465259 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15465259 Supplier Thermo Scientific Alfa Aesar Supplier No. J62969.03
  • ONLINE EXCLUSIVE PRICE
    £74.10
    £60.65 / 1g
    Save £13.45
    18% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 30484-77-6 | C26H28Cl2F2N2 | 477.42 g/mol

A dual sodium/calcium channel blocker; a cerebral and peripheral vasodilator

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 30484-77-6
Color White
Molecular Formula C26H28Cl2F2N2
MDL Number MFCD00058198
Quantity 1 g
Beilstein 4284243
Merck Index 14,4144
Synonym flunarizine dihydrochloride, flunarizine hydrochloride, flunarizine 2hcl, flunarizine hcl, flunarizine hydrochloride usan:jan, 1-bis 4-fluorophenyl methyl-4-cinnamylpiperazine dihydrochloride, e-1-bis-p-fluorophenyl methyl-4-cinnamylpiperazine dihydrochloride, e-1-bis 4-fluorophenyl methyl-4-3-phenyl-2-propenyl piperazine dihydrochloride, e-1-bis 4-fluorophenyl methyl-4-cinnamylpiperazine dihydrochloride
InChI Key RXKMOPXNWTYEHI-RDRKJGRWSA-N
SMILES [H+].[H+].[Cl-].[Cl-].FC1=CC=C(C=C1)C(N1CCN(C\C=C\C2=CC=CC=C2)CC1)C1=CC=C(F)C=C1
IUPAC Name dihydrogen 1-[bis(4-fluorophenyl)methyl]-4-[(2E)-3-phenylprop-2-en-1-yl]piperazine dichloride
Molecular Weight (g/mol) 477.42
PubChem CID 5282407
Formula Weight 477.4
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Flunarizine dihydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.