missing translation for 'onlineSavingsMsg'
Learn More

Flunarizine Dihydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30460622
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460622 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460622 Supplier LoGiCal Supplier No. LGCAMP0175.0001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Flunarizine Dihydrochloride 1.0 mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flunarizine dihydrochloride
CAS 30484-77-6
Analyte or Component Names Flunarizine dihydrochloride
Shipping Condition Ice Pack (-20°C)
Molecular Formula C26 H26 F2 N2 . 2 Cl H
Certifications/Compliance ISO 17034
Synonym Piperazine, 1-[bis(4-fluorophenyl)methyl]-4-[(2 E)-3-phenyl-2-propen-1-yl]-, hydrochloride (1:2), Piperazine, 1-[bis(4-fluorophenyl)methyl]-4-(3-phenyl-2-propenyl)-, dihydrochloride, (E)-, Piperazine, 1-[bis(4-fluorophenyl)methyl]-4-[(2 E)-3-phenyl-2-propenyl]-, dihydrochloride (9 CI), Piperazine, 1-[bis(p-fluorophenyl)methyl]-4-cinnamyl-, dihydrochloride, (E)- (8 CI), (E)-1-[Bis(4-fluorophenyl)methyl]-4-cinnamylpiperazine dihydrochloride, Dinaplex, Flugeral, Flunagen, Flunarizine dihydrochloride, Flunarizine hydrochloride, Flunarl, Fluxarten, Gradient, Issium, Mondus, R 14950
InChI Formula InChI=1 S/C26H26F2N2.2 ClH/c27-24-12-8-22(9-13-24)26(23-10-14-25(28)15-11-23)30-19-17-29(18-20-30)16-4-7-21-5-2-1-3-6-21;;/h1-15,26 H,16-20H2;2*1 H/b7-4+;;
SMILES Cl.Cl.Fc1ccc(cc1)C(N2CCN(C\C=C\c3ccccc3)CC2)c4ccc(F)cc4
IUPAC Name 1-[bis(4-fluorophenyl)methyl]-4-[(E)-3-phenylprop-2-enyl]piperazine;dihydrochloride
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 477.42
Quantity 1 mL
Formula Weight 476.1598 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.