missing translation for 'onlineSavingsMsg'
Learn More

Flumorph, TRC

Product Code. 30398036
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398036 10 mg -
30389093 25 mg -
30404928 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398036 Supplier Toronto Research Chemicals Supplier No. TRCF45502510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 211867-47-9
Molecular Formula C21 H22 F N O4
Molecular Weight (g/mol) 371.4
Synonym Morpholine, 4-[3-(3,4-dimethoxyphenyl)-3-(4-fluorophenyl)-1-oxo-2-propenyl]- (9CI), 3-(3,4-Dimethoxyphenyl)-3-(4-fluorophenyl)-1-(4-morpholinyl)-2-propen-1-one, (EZ)-3-(3,4-dimethoxyphenyl)-3-(4-fluorophenyl)-1-morpholinopropenone, (EZ)-4-[3-(3,4-dimethoxyphenyl)-3-(4-fluorophenyl)acryloyl]morpholine, 4-[3-(3,4-Dimethoxyphenyl)-3-(4-fluorophenyl)-1-oxo-2-propenyl]morpholine, 4-[3-(4-Fluorophenyl)-3-(3,4-dimethoxyphenyl)acryloyl]morpholine, Flumorph, Fumalin, SYP-L 190
IUPAC Name 3-(3,4-dimethoxyphenyl)-3-(4-fluorophenyl)-1-morpholin-4-ylprop-2-en-1-one
SMILES COc1ccc(cc1OC)C(=CC(=O)N2CCOCC2)c3ccc(F)cc3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H22FNO4/c1-25-19-8-5-16(13-20(19)26-2)18(15-3-6-17(22)7-4-15)14-21(24)23-9-11-27-12-10-23/h3-8,13-14H,9-12H2,1-2H3
Formula Weight 371.1533
Chemical Name or Material Flumorph
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.