missing translation for 'onlineSavingsMsg'
Learn More

Flumequine 13C3 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30321944
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321944 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321944 Supplier Dr. Ehrenstorfer Supplier No. DREA13718010AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flumequine-13C3
CAS 1185049-09-5
Solvent or Matrix Acetonitrile
Molecular Formula 13C3 C11 H12 F N O3
Quantity 1 mL
InChI Formula InChI=1S/C14H12FNO3/c1-7-2-3-8-4-9(15)5-10-12(8)16(7)6-11(13(10)17)14(18)19/h4-7H,2-3H2,1H3,(H,18,19)/i11+1,13+1,14+1
SMILES CC1CCc2cc(F)cc3[13C](=O)[13C](=CN1c23)[13C](=O)O
Molecular Weight (g/mol) 264.23
Formula Weight 264.0902 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.