missing translation for 'onlineSavingsMsg'
Learn More

Flumazenil, TRC

Product Code. 30406187
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30406187 50 mg -
30401409 10 mg 10mg
30394887 100 mg -
3 options
This item is not returnable. View return policy

Product Code. 30406187

Brand: Toronto Research Chemicals TRCF50045050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78755-81-4
Formula Weight 303.1019
InChI Formula InChI=1S/C15H14FN3O3/c1-3-22-15(21)13-12-7-18(2)14(20)10-6-9(16)4-5-11(10)19(12)8-17-13/h4-6,8H,3,7H2,1-2H3
IUPAC Name ethyl 8-fluoro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
Molecular Formula C15 H14 F N3 O3
Molecular Weight (g/mol) 303.29
Percent Purity >95%
SMILES CCOC(=O)c1ncn2c1CN(C)C(=O)c3cc(F)ccc23
Synonym Flumazenil, 4H-Imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic acid, 8-fluoro-5,6-dihydro-5-methyl-6-oxo-, ethyl ester, 8-Fluoro-5,6-dihydro-5-methyl-6-oxo-4H-Imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic acid ethyl ester, Ethyl 8-fluoro-5-methyl-6-oxo-5,6-dihydro-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate, Anexate, Flumazepil, Flumenazil, Lanexat, Mazicon, Ro 15-1788, Ro 15-1788/000, Ro 151788, Ro 1722, Ro 41-8157, Romazicon
Chemical Name or Material Flumazenil
Grade HPLC
Show More Show Less

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.