missing translation for 'onlineSavingsMsg'
Learn More

Flumazenil, TRC

Product Code. 30394887
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394887 100 mg -
30406187 50 mg -
30401409 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394887 Supplier Toronto Research Chemicals Supplier No. TRCF500450100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78755-81-4
Molecular Formula C15 H14 F N3 O3
Molecular Weight (g/mol) 303.29
Synonym Flumazenil, 4H-Imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic acid, 8-fluoro-5,6-dihydro-5-methyl-6-oxo-, ethyl ester, 8-Fluoro-5,6-dihydro-5-methyl-6-oxo-4H-Imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic acid ethyl ester, Ethyl 8-fluoro-5-methyl-6-oxo-5,6-dihydro-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate, Anexate, Flumazepil, Flumenazil, Lanexat, Mazicon, Ro 15-1788, Ro 15-1788/000, Ro 151788, Ro 1722, Ro 41-8157, Romazicon
IUPAC Name ethyl 8-fluoro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
SMILES CCOC(=O)c1ncn2c1CN(C)C(=O)c3cc(F)ccc23

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H14FN3O3/c1-3-22-15(21)13-12-7-18(2)14(20)10-6-9(16)4-5-11(10)19(12)8-17-13/h4-6,8H,3,7H2,1-2H3
Formula Weight 303.1019
Percent Purity >95%
Chemical Name or Material Flumazenil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.