missing translation for 'onlineSavingsMsg'
Learn More

Flumazenil, MedChemExpress

Product Code. 16488056
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488056 100 mg 100mg
16424207 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488056 Supplier Medchem Express Supplier No. HYB0009
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Flumazenil is a competitive GABAA receptor antagonist, used in the treatment of benzodiazepine overdoses.

Flumazenil is a competitive GABAA receptor antagonist, used in the treatment of benzodiazepine overdoses.

Specifications

Chemical Name or Material Flumazenil
CAS 78755-81-4
Color White
Purity Grade Notes Research
Molecular Formula C15H14FN3O3
Quantity 100 mg
Synonym Ro 15-1788
Solubility Information DMSO : 20 mg/mL (65.94 mM; Need ultrasonic)
SMILES O=C(C1=C2CN(C(C3=CC(F)=CC=C3N2C=N1)=O)C)OCC
Molecular Weight (g/mol) 303.29
Formula Weight 303.29
Percent Purity 98.9%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.