missing translation for 'onlineSavingsMsg'
Learn More

Flumazenil, LoGiCal

Product Code. 30457481
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457481 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457481 Supplier LoGiCal Supplier No. LGCFOR0762.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Flumazenil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flumazenil
CAS 78755-81-4
Analyte or Component Names Flumazenil
Shipping Condition Room Temperature
Molecular Formula C15 H14 F N3 O3
Certifications/Compliance ISO 9001
Synonym Flumazenil, Ethyl 8-fluoro-5-methyl-6-oxo-5,6-dihydro-4 H-imidazo[1,5-a][1,4 ]benzodiazepine-3-carboxylate, Anexate, Flumazenil, Flumazepil, Flumenazil, Lanexat, Mazicon, Ro 15-1788, Ro 15-1788/000, Ro 151788, Ro 1722, Ro 41-8157, Romazicon
InChI Formula InChI=1 S/C15H14FN3O3/c1-3-22-15(21)13-12-7-18(2)14(20)10-6-9(16)4-5-11(10)19(12)8-17-13/h4-6,8 H,3,7H2,1-2H3
SMILES CCOC(=O)c1ncn2c1CN(C)C(=O)c3cc(F)ccc23
IUPAC Name ethyl 8-fluoro-5-methyl-6-oxo-4 H-imidazo[1,5-a][1,4 ]benzodiazepine-3-carboxylate
Industry Type Forensics
Molecular Weight (g/mol) 303.29
Quantity 10 mg
Formula Weight 303.1019 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.