missing translation for 'onlineSavingsMsg'
Learn More

Flumazenil, 98%

Product Code. 15494899
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494899 100 mg 100mg
15404909 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494899 Supplier Thermo Scientific Alfa Aesar Supplier No. J62537.MC
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Benzodiazepine antagonist | CAS: 78755-81-4 | C15H14FN3O3 | 303.293 g/mol

Benzodiazepine antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78755-81-4
Molecular Formula C15H14FN3O3
Molecular Weight (g/mol) 303.293
MDL Number MFCD00242764
InChI Key OFBIFZUFASYYRE-UHFFFAOYSA-N
Synonym flumazenil, anexate, flumazepil, romazicon, lanexat, mazicon, flumazenilum, flumazenilo, flumazenilum latin, flumazenilo spanish
PubChem CID 3373
ChEBI CHEBI:5103
IUPAC Name ethyl 8-fluoro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
SMILES CCOC(=O)C1=C2CN(C(=O)C3=C(N2C=N1)C=CC(=C3)F)C

Specifications

Color White
Quantity 100 mg
Merck Index 14,4135
Solubility Information Soluble in DMSO to 25mM
Formula Weight 303.29
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Flumazenil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.