missing translation for 'onlineSavingsMsg'
Learn More

Flumazenil, 98%, Thermo Scientific™

Benzodiazepine antagonist

Brand:  Thermo Scientific Alfa Aesar J62537.ME

Quantity 500mg

 View more versions of this product

Product Code. 15404909

  • £851.00 / 500mg
 Restricted Item
Estimated Shipment: 17-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

78755-81-4
303.293
OFBIFZUFASYYRE-UHFFFAOYSA-N
3373
ethyl 8-fluoro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
C15H14FN3O3
MFCD00242764
flumazenil, anexate, flumazepil, romazicon, lanexat, mazicon, flumazenilum, flumazenilo, flumazenilum latin, flumazenilo spanish
CHEBI:5103
CCOC(=O)C1=C2CN(C(=O)C3=C(N2C=N1)C=CC(=C3)F)C

Specifications

78755-81-4
MFCD00242764
14,4135
Soluble in DMSO to 25mM
CCOC(=O)C1=C2CN(C(=O)C3=C(N2C=N1)C=CC(=C3)F)C
303.293
CHEBI:5103
98%
C15H14FN3O3
500mg
flumazenil, anexate, flumazepil, romazicon, lanexat, mazicon, flumazenilum, flumazenilo, flumazenilum latin, flumazenilo spanish
OFBIFZUFASYYRE-UHFFFAOYSA-N
ethyl 8-fluoro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
3373
303.29
Flumazenil
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.