Learn More
HALYARD™ FLUIDSHIELD™ Procedure Masks, Type IIR
Product Code. 30274763 Shop All Halyard Products
Change view
Click to view available options
Color:
Blue
Orange
Includes:
No visor
Visor
Unit Size:
Pack of 100
Pack of 400
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Includes unitSize
30274763 Orange No visor Pack of 400
30274711 Blue No visor Pack of 500
30274766 Blue Visor Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30274763 Supplier HALYARD™ Supplier No. 47107
Procedure Masks provide extra protection for procedures where fluid splashes are possible.
  • Pleated, 4-ply facemask, with elastic earloops
  • Type IIR mask as defined by EN 14683:2019+AC:2019
  • Medical Device class I per Regulation (EU) 2017/745
  • Made of Polypropylene and Cellulose/Polyester nonwovens
  • Available with a splash resistance of 120 (FLUIDSHIELD Level 2) or 160 mm Hg (FLUIDSHIELD Level 3), tested per ASTM F2100-11
  • Model 62116 with wraparound visor for eye protection and with Fog-free foam band to avoid fogging
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 3
Color Orange
Fastening Type Earloops
Material Spunbond Polypropylene
Size One Size
Sterility Non-sterile
Style Pleated
Certifications/Compliance Medical Device class I per Regulation (EU) 2017/745, Type IIR per EN 14683:2019+AC:2019, Level 3 per ASTM F2100-11
Disposable Single-use
Includes No visor
Latex Free Yes