missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FluidEase™ Pro Finnpipette™ Electronic Single-Channel Pipettes

Product Code. 32108091
Change view
Click to view available options
Volume (Metric):
0.25 to 5 mL
0.5 to 10 μL
0.5 to 10 mL
10 to 300 μL
2.5 to 100 μL
20 to 1000 μL
5 to 200 μL
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
32108091 5 to 200 μL Each
32108101 0.5 to 10 μL Each
32108104 2.5 to 100 μL Each
32108099 10 to 300 μL Each
32108095 20 to 1000 μL Each
32108097 0.25 to 5 mL Each
32108090 0.5 to 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 32108091 Supplier Thermo Scientific™ Supplier No. 4673030
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ FluidEase™ Pro Finnpipette™ electronic single-channel pipettes deliver precise, reproducible liquid handling with greater tip choice, intuitive touchscreen operation, and ergonomic comfort for productive everyday pipetting.

Thermo Scientific™ FluidEase™ Pro Finnpipette™ electronic single-channel pipettes deliver reliable, reproducible liquid handling with greater tip choice. A large, intuitive touchscreen, ultra-wide speed selection, ergonomic design, and flexible programming simplify workflows, while password protection and service and calibration logs support standardization and confident operation in shared and regulated laboratories.

  • Large Touchscreen User Interface: A high-resolution touchscreen provides direct, visual interaction for setup and execution, reducing training time and minimizing setup errors compared to button- or joystick-based navigation.
  • Ultra-Wide Speed Selection: One of the broadest aspiration and dispense speed ranges available supports gentle handling of sensitive samples, precise control of viscous liquids, and faster execution for time-critical workflows.
  • Simple Presets and Advanced Programming: Easy-to-use preset modes support fast onboarding for new or occasional users, while advanced step programming enables complex, multi-step protocols for demanding applications.
  • Index-Finger Operation with Increased Rotational Flexibility: Retains Thermo Fisher–recognized index-finger operation with a wider angle of rotation at the finger rest to accommodate different hand sizes, grip angles, and working positions.
  • Wide Selection of Finntip™ Pipette Tips: Thermo Scientific™ Finntip™ pipette tips are designed and manufactured for Finnpipette™ models to support optimal performance, precision, and accuracy, with a wide selection of tips for diverse applications and workflows.
  • Electronic Tip Ejection with Safe-to-Eject Illumination: Electronic tip ejection reduces physical effort across the range, including 5 mL and 10 mL pipette models, while LED illumination indicates when tip ejection is possible for added confidence and ergonomic comfort.
  • Password Protection and User Profiles: Password-protect critical settings and protocols and support up to five user profiles to enable standardized operation in shared and regulated laboratory environments.
  • Custom Calibration Options: Enable calibration to customer-specific liquids or environmental conditions to align performance with real laboratory workflows.
  • Service and Maintenance Tracking: Built-in service reminders, calibration logs, and usage tracking support instrument management and compliance documentation over time.
TRUSTED_SUSTAINABILITY

Specifications

Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Yellow
Disposable Reusable
For Use With FinnTip 200, Flex 200, 200
Model Bluetooth
Number of Channels 1
Programmability Yes
Sterility Non-sterile
Type Electronic Single Channel Pipette
Volume (Metric) 5 to 200 μL
Display Touchscreen
Finger Style Ergonomic Index Finger
Increments 0.1 μL
Multilingual User Interface Yes
Warranty 1 Year, 3 Years with Registration
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.