Learn More
Thermo Scientific™ Fluid Transfer Assembly
Specialty Fluid Transfer Assembly products integrate easily within your existing process and equipment to maximize productivity, reduce cross-contamination and keep your process running smoothly.
Brand: Thermo Scientific™ SS00063-I
This item is not returnable.
View return policy
Description
Specialized connectors and tubing in our ready-designed assemblies provide an off-the-shelf solution that addresses many fluid transfer needs while delivering efficiency and increasing process integrity.- End treatments facilitate simple interconnection with existing process and equipment
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
Specifications
| 91.4cm of 12.7cm ID Silicone Tubing with 1.9cm Sanitary and 1.3cm MPX Body Connections |