missing translation for 'onlineSavingsMsg'
Learn More

Flufenoxuron, TRC

Product Code. 30396283
Change view
Click to view available options
Quantity:
500 mg
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396283 10 g
30408583 25 g
30398437 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396283 Supplier Toronto Research Chemicals Supplier No. TRCF43510010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101463-69-8
Molecular Formula C21 H11 Cl F6 N2 O3
Molecular Weight (g/mol) 488.77
Synonym N-[[[4-[2-Chloro-4-(trifluoromethyl)phenoxy]-2-fluorophenyl]amino]carbonyl]-2,6-difluorobenzamide, Cascade, Cascade (pesticide), Flufenoxuron, WL 115110, Benzamide, N-[[[4-[2-chloro-4-(trifluoromethyl)phenoxy]-2-fluorophenyl]amino]carbonyl]-2,6-difluoro-
IUPAC Name N-[[4-[2-chloro-4-(trifluoromethyl)phenoxy]-2-fluorophenyl]carbamoyl]-2,6-difluorobenzamide
SMILES Fc1cccc(F)c1C(=O)NC(=O)Nc2ccc(Oc3ccc(cc3Cl)C(F)(F)F)cc2F

Specifications

Quantity 10 g
InChI Formula InChI=1S/C21H11ClF6N2O3/c22-12-8-10(21(26,27)28)4-7-17(12)33-11-5-6-16(15(25)9-11)29-20(32)30-19(31)18-13(23)2-1-3-14(18)24/h1-9H,(H2,29,30,31,32)
Formula Weight 488.0362
Chemical Name or Material Flufenoxuron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.