missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Flufenamic acid, 97%

Product Code. 10144420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10144420 10 g Glass bottle 10g
10713551 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10144420 Supplier Thermo Scientific Chemicals Supplier No. 165920100
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 530-78-9 | C14H10F3NO2 | 281.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-78-9
Molecular Formula C14H10F3NO2
Molecular Weight (g/mol) 281.22
MDL Number MFCD00002422
InChI Key LPEPZBJOKDYZAD-UHFFFAOYSA-N
Synonym flufenamic acid, fluphenamic acid, nichisedan, achless, arlef, flufacid, fullsafe, lanceat, paraflu, plostene
PubChem CID 3371
ChEBI CHEBI:42638
IUPAC Name 2-[3-(trifluoromethyl)anilino]benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)NC2=CC=CC(=C2)C(F)(F)F

Specifications

Melting Point 132°C to 135°C
Color Gray-Green to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula 2-(CF3C6H4NH)C6H4CO2H
Quantity 10 g
Beilstein 14, III, 905
Merck Index 15, 4163
Solubility Information Solubility in water: 0.0265g/L (37°C). Other solubilities: soluble in ethanol (6.9g/L at 37°C),chloroform,(19g/L at 37°C),dmso,cyclohexane
Formula Weight 281.22
Percent Purity 97%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Flufenamic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Flufenamic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.