missing translation for 'onlineSavingsMsg'
Learn More

Fludrocortisone Acetate, TRC

Product Code. 30389434
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389434 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389434 Supplier Toronto Research Chemicals Supplier No. TRCF4281011G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fludrocortisone Acetate
CAS 514-36-3
Molecular Formula C23 H31 F O6
Quantity 1 g
Synonym Fludrocortisone acetate, 9-Fluoro-11β,17-dihydroxy-3,20-dioxopregn-4-en-21-yl acetate, Corticosterone, 9-fluoro-17-hydroxy-, 21-acetate (6CI), Pregn-4-ene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-, 21-acetate (7CI,8CI), 9-Fluorohydrocortisone acetate, 9α-Fluoro-11β,17α,21-trihydroxypregn-4-ene-3,20-dione 21-acetate, 9α-Fluorocortisol 21-acetate, 9α-Fluorocortisol acetate, 9α-Fluorohydrocortisone 21-acetate, 9α-Fluorohydrocortisone acetate, Alflorone acetate, Cortineff, Fludrocortisone 21-acetate, Fludrocortisone acetate, Fluorocortisol acetate, Fluorohydrocortisone acetate, NSC 15186, Scherofluron, U 4845
InChI Formula InChI=1S/C23H31FO6/c1-13(25)30-12-19(28)22(29)9-7-16-17-5-4-14-10-15(26)6-8-20(14,2)23(17,24)18(27)11-21(16,22)3/h10,16-18,27,29H,4-9,11-12H2,1-3H3/t16-,17-,18-,20-,21-,22-,23-/m0/s1
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
IUPAC Name [2-[(8S,9R,10S,11S,13S,14S,17R)-9-fluoro-11,17-dihydroxy-10,13-dimethyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molecular Weight (g/mol) 422.49
Formula Weight 422.2105
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.