missing translation for 'onlineSavingsMsg'
Learn More

Fludarabine phosphate, MedChemExpress

Product Code. 16478066
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478066 100 mg 100mg
16458328 10 mg 10mg
16414217 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478066 Supplier Medchem Express Supplier No. HYB0028
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fludarabine (phosphate) is an analogue of adenosine and deoxyadenosine, which is able to compete with dATP for incorporation into DNA and inhibit DNA synthesis.

Fludarabine (phosphate) is an analogue of adenosine and deoxyadenosine, which is able to compete with dATP for incorporation into DNA and inhibit DNA synthesis.

Specifications

Chemical Name or Material Fludarabine phosphate
CAS 75607-67-9
Color White
Purity Grade Notes Research
Molecular Formula C10H13FN5O7P
Quantity 100 mg
Synonym NSC 118218 phosphate
Solubility Information DMSO : ≥ 100 mg/mL (273.82 mM) ∣H2O : 5 mg/mL (13.69 mM; Need ultrasonic)
SMILES NC1=C2C(N([C@H]3[C@@H](O)[C@H](O)[C@@H](COP(O)(O)=O)O3)C=N2)=NC(F)=N1
Molecular Weight (g/mol) 365.21
Formula Weight 365.21
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.