missing translation for 'onlineSavingsMsg'
Learn More

Flucytosine, Mikromol™

Product Code. 17962526
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962526 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17962526 Supplier Mikromol Supplier No. MM0355.00
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Flucytosine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Fluorocytosine
CAS 2022-85-7
Analyte or Component Names 5-Fluorocytosine
Shipping Condition Room Temperature
Molecular Formula C4 H4 F N3 O
Certifications/Compliance ISO 17025
Synonym Flucytosine
InChI Formula InChI=1S/C4H4FN3O/c5-2-1-7-4(9)8-3(2)6/h1H,(H3,6,7,8,9)
SMILES NC1=NC(=O)NC=C1F
IUPAC Name 4-amino-5-fluoro-1H-pyrimidin-2-one
Molecular Weight (g/mol) 129.09
Quantity 250 mg
Formula Weight 129.0338 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.