missing translation for 'onlineSavingsMsg'
Learn More

Fluconazole, MedChemExpress

Product Code. 16488086
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488086 100 mg 100mg
16422016 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488086 Supplier Medchem Express Supplier No. HYB0101
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fluconazole (UK-49858) is a triazole antifungal agent with excellent activities against a broad range of fungi, especially against Candida albicans. Fluconazole inhibits C. albicans and Candida kefyr with IC99s range from 0.20 μg/mL to 0.39 μg/mL.

Fluconazole (UK-49858) is a triazole antifungal agent with excellent activities against a broad range of fungi, especially against Candida albicans. Fluconazole inhibits C. albicans and Candida kefyr with IC99s range from 0.20 μg/mL to 0.39 μg/mL.

Specifications

Chemical Name or Material Fluconazole
Physical Form Powder
CAS 86386-73-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H12F2N6O
For Use With (Application) COVID-19-immunoregulation
Synonym UK-49858
Solubility Information DMSO : ≥ 100 mg/mL (326.51 mM) ∣H2O : 2 mg/mL (6.53 mM; Need ultrasonic)
SMILES FC1=CC=C(C(O)(CN2C=NC=N2)CN3C=NC=N3)C(F)=C1
Molecular Weight (g/mol) 306.27
Formula Weight 306.27
Percent Purity 99.21%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.